Stainless Steel Manifolds for Microtest Plates, Drummond
Supplier: Drummond
3-000-093 3-000-094 3-000-096 3-000-097 3-000-098 3-000-099
53504-228EA 565.81 CAD
53504-228 53504-239 53504-240 53504-250 53504-261 53507-366
Stainless Steel Manifolds for Microtest Plates, Drummond
Manifolds
Stainless steel/PTFE manifolds allow multiple test positions to be fed, rinsed, or aspirated quickly and easily.
  • Autoclavable
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection
  • Pin has O.D. 0.031" and I.D. 0.018"
The autoclavable manifolds work for both sample delivery and aspiration operations. They include a cleaning wire for easy maintenance.