Wiretrol® II Disposable Micropipettes, Drummond
Supplier: Drummond
5-000-2200 5-000-2005 5-000-2010 5-000-2020 5-000-2050 5-000-2100
53480-655EA 137.35 CAD
53480-655 53507-426 53507-428 53507-430 53507-432 53507-434
Wiretrol® II Disposable Micropipettes, Drummond
Pipettes Microcapillaries
This economical positive-displacement micropipette features fingertip control.
  • Dual graduations for non-contact delivery
  • Ideal for a wide variety of applications including both high- and low-viscosity liquids and corrosives
  • Economical
The Wiretrol II provides the accuracy and convenience of the Wiretrol I series with dual graduations and a shorter plunger for non-contact delivery.
Order Now
SPECIFICATIONS
Environmentally Preferable