Sampling Tubes and Systems, Drager
Supplier: Draeger
8101821 6728631 6728891 6728811
CA13453-468PK 529.39 CAD
CA13453-468 CA13453-470 CA34330-184 13453-472
Sampling Tubes and Systems, Drager
Gas Detection Systems
Using the Drager sampling system, hazardous substances in the air are collected using a suitable medium via adsorption or chemisorption.
In active sampling, the air to be evaluated is drawn through a sampling tube such as Activated Charcoal and Silica Gel tube types with a pump and passive sampling is performed with diffusion samplers like the Dräger ORSA or the Dräger Nitrous Oxide diffusion samplers.