Simulaids® Choking Training Torsos
Supplier: Nasco
100-1602 100-1615 100-1620
470119-774EA 1115.95 CAD
470119-774 470119-776 470119-778
Simulaids® Choking Training Torsos
Models Human Biology Models
Practice Emergency Choking Procedures
  • Can be used for heimlich and back blow procedures
  • Proper procedure is rewarded with expiation of the blockage
  • Includes carrying bag
This life-size torso allows practice of abdominal/chest thrust back blow procedures (Heimlich maneuver) and mouth sweep for clearing a blocked airway. When correct procedures are performed, the manikin will expel the object causing an obstruction. Made of durable vinyl to create tactile realism. Features include: Anatomical landmarks such as ribcage, xiphoid process and jugular notch. Supplied with choking objects, shirt and soft carrying bag.