Simulaids® Adult CPR Manikin With Electronics
Supplier: Nasco
100-2725 100-2507 150-1374
470119-796EA 4031.2 CAD
470119-796 470119-802 470119-804 470119-960
Simulaids® Adult CPR Manikin With Electronics
Models Human Biology Models
Full Sized Adult Manikin
  • Easy to clean
  • Replaceable faces and airways
  • Easily positionable
  • A carry bag is available
This full-size manikin allows the training of BLS rescue techniques and CPR. The resiliency and weight of the body is very realistic. Features include: Natural resistance to chest compression, Palpable carotid pulse, Anatomical landmarks such as sternum, ribcage and sub-sternal notch, Airway ball valve which allows the lungs to inflate only if the head is extended and accurately positioned, Light controller which confirms correct handMore Product Information