Simulaids® CPR Torso
Supplier: Simulaids
100-2000 100-2805
470119-828EA 201.9 CAD
470119-828 470119-832
Simulaids® CPR Torso
First Aid Supplies CPR Products
Economical Adult CPR Training.
  • Easy to clean
  • Replaceable faces and airways
  • Includes soft carry bag
his economical CPR manikin is constructed of soft, realistic vinyl plastic over polyurethane foam for a 'human' feel. Features include: a longer torso for realistic abdominal thrusts, realistic head tilt and chin lift for opening airway, can easily be manipulated to realistically simulate airway obstruction or choking situations, and a user-friendly lung/airway design that eliminates cleaning. Includes three mouth/nose pieces and three disposable lung/airway systems. Nylon carry bag and kneeling pads included.