Life/form® CRiSis™ Training Manikins
Supplier: Nasco
LF03215 LF03958
470119-340EA 488.9 CAD
470119-340 470119-898 470119-904
Life/form® CRiSis™ Training Manikins
Educational Materials Biology Educational Materials Human Biology Learning Activities
Ideal For Acls, Paramedic, Emt, And Nursing Training At Every Level
  • Useable with many defibrillatiors
  • Controllable blood pressure arm
  • Realistic anatomy for airway management
The Deluxe “PLUS” Adult CRiSis™ Manikin is a complete resuscitation system that presents challenges you may face in real world situations, with five major features allowing practice of several simulation scenarios. Each manikin consists of an “Airway Larry” Airway Management Trainer Head, Blood Pressure Arm, Defibrillation Chest Skin, full body CPR manikin, and IV Arm. Ideal for ACLS, paramedic, EMT, and nursing training at every level. Modular components allow you to create a manikin to suit your needs.
The “PLUS” manikin also includes an Electronic CPR Monitoring Printer Unit and a Hard Carry Case. Electronic monitoring, memory, and printer unit, for use with CPR simulation, displays running averages of compression rate, compression depth, ventilation duration, and ventilation volume; provides hard copy printout; and has adult and child settings. Red light indicates improper hand placement, yellow light signals proper compression depth of 1-1/2""-2"". Green light signals proper ventilation volume at 0.80 liters adult. The performance of each skill is displayed separately while averages are stored in the memory. Designed to automatically switch between the compression and ventilation modes depending on which one is being applied. Compression and ventilation waveforms are plotted in a time sequence. When stop button is pushed, averages of the four skills are shown on the digital display while the printer tabulates the result on paper.
Life/form® Deluxe CRiSis™ Torso Manikin features the same quality, durability, and reliability as the Deluxe CRiSis™ Manikin only in a smaller, easier-to-transport version. Unlike full-body manikins, this ACLS torso is much smaller and easier to move from facility to facility for training. This system now features the “Airway Larry” Airway Management Training Head with carotid pulse, Defibrillation Chest Skin, and Interactive ECG Simulator , all attached to a CPARLENE® Torso. Includes a 8-oz. pump spray bottle of lubricant and comes in a durable, foam-lined carton.