Life/form® Pediatric Head Injection Trainer
Supplier: Nasco
LF00999 LF01003 1005632
470119-836EA 1585.6 CAD
470119-836 470119-838 470119-840
Life/form® Pediatric Head Injection Trainer
Models Human Biology Models
Practice Temporal And Jugular Infusions And Injections
  • Realistically modeled on a 6 month old
  • Life like skin allows palpation for the vein
  • Included carrying case
An invaluable tool for demonstrating and practicing infusions and injections in the temporal and jugular veins of a newborn to a twelve-month-old infant. The neck is made of soft, flexible foam to provide a realistic feel of palpation and puncture. The pediatric head includes the following: Head (to represent a six-month-old infant) with skin and veins, Fluid supply bag, 2 different gaugeMore Product Information