Life/form® Enema Administration Trainer
Supplier: Nasco
LF00957 LF01050
470119-382EA 1999.25 CAD
470119-382 470119-384
Life/form® Enema Administration Trainer
Models Human Biology Models
Good For Demonstrations And Practice Sessions
  • Flexible buttock for realism
  • No special equipment required
  • Includes carrying case
All aspects of the simulator are lifelike in every detail. The upper buttock is flexible so that the trainee must raise the buttock in order to locate and make insertion into the anus for a lifelike enema. Portable and easy to keep clean, the simulator features a 1 liter fluid reservoir which is adequate for most demonstration and practice purposes. For large groups, a drainage tube is provided so that the fluid can beMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable