Life/form® Arterial Puncture Arm
Supplier: Nasco
LF00995 1005605
470119-412EA 1980.45 CAD
470119-412 470119-414
Life/form® Arterial Puncture Arm
Models Human Biology Models
Easy To Set Up And Use
  • Replicable skin and arteries
  • Includes tubing sealant for extended tubing usage
  • Includes carrying case
This easy to use training aid is ideal for practice in and demonstration of drawing arterial blood samples and for monitoring blood gases.
Puncture locations can be identified through palpation of the pulse of radial and brachial artery. Realistic arterial pressure produces a lifelike backflow of blood in the syringe, confirming proper needle location in the artery.
Delivered with 2 syringes and tubules, artificial arterial blood, 2 replacement arterial sections and carrying case.