Dricyclers, Boekel Scientific
Supplier: Boekel
134041 134441
13278-904EA 6485.08 CAD
13278-904 CA70380-002
Dricyclers, Boekel Scientific
Desiccators Desiccator Cabinets
The Boekel Scientific series of Dricyclers are perfect for storing reagents or soil samples, drying biological specimens, cooling crucibles or the storage of sludge samples. These Dricyclers automatically regenerate the silica gel desiccant enclosed in the control unit thereby eliminating the need to replace or bake desiccant. In the drying sequence, the Dricyclers automatically regenerate desiccant every five hours. The silica gel cassette housed in the control box is exposed to desiccator air for 4.5 hours and then is closed off from the desiccator air and regenerated by heating. The moisture is vented to the outside air.
  • Durable hand crafted construction
  • Fabricated from stainless steel and glass
  • Shelves can be adjusted in half-inch increments
  • Superior seal integrity with neoprene gasket
  • Automatic desiccant regeneration
  • Easy to read hygrometer
Available in a small or large size and choose your shelving type, based on your application. The large unit is available only with two stainless steel shelves. Also available in a 230 V model.
Certifications: CE compliant.
Ordering information: All Boekel Scientific Dricyclers come equipped with a door mounted hygrometer, two shelves and a two year manufacturers warranty.