GlycanPac AXR-1 Glycan Isomer Analysis Columns, Thermo Scientific
Supplier: Thermo Scientific
088135 088136
10148-468EA 2142.24 CAD
10148-468 10148-470 10148-498 10148-500 10148-524 10148-526 10148-528 10148-530
GlycanPac AXR-1 Glycan Isomer Analysis Columns, Thermo Scientific
Chromatography Columns
The Thermo Scientific™ GlycanPac™ AXR-1 column is a high-performance, silica-based HPLC column for the simultaneous separation of glycans by charge, isomerism, and size. It is designed to provide industry-leading resolution with unique selectivities for biologically important glycans, either labeled or native, using either fluorescence and/or mass spectrometry (MS) detection.
  • Unique glycan selectivity based on charge, branch/linkage isomerism, and size
  • Exceptional resolution for either native or derivatized glycans
  • Compatibility with both MS and
More Product Information