VWR® Tube Rocker Collection
Supplier: VWR International
10159-752 10159-754 10159-756
10159-752EA 1123.54 CAD
10159-752 10159-754 10159-756
VWR® Tube Rocker Collection
Shakers Rotating/Rocking Shakers
The VWR® Tube Rocker Collection features Tube Rockers available in a "Mini" size as well as an "XL" size and in Fixed Speed or a Variable Speed models.
  • Low profile and small footprint
  • Tube Rockers provide smooth, gentle mixing
  • Available with a variety of speed settings
The Mini Tube Rockers holds 8 tubes, 120 mm long while the XL model holds up to 18 tubes, also 120 mm long. Each model is equipped with a fixed tilt angle of 35 degrees and mixes your samples simultaneously via a 110-240V power pack. The variable speed models are adjustable in 1 rpmMore Product Information