Life/form® Prostate Exam Simulator
Supplier: Nasco
LF00901
470119-394EA 3110.65 CAD
470119-394
Life/form® Prostate Exam Simulator
Models Human Biology Models
Teach Prostate Exam Without Real Patients
  • Includes 4 different prostate inserts
  • Includes guide and case
This prostate exam simulator consists of a male abdomen body and 4 different interchangeable prostate glands which can be inserted to allow realistic practice in diagnosis of prostate cancer by rectal examination.
The 4 prostate glands represent the following characteristics:
Benign, slightly enlarged, but otherwise normal prostate
Beginning stage of carcinoma, a discrete, hard nodule is palpable in the upper right quadrant of prostate
The spread ofMore Product Information