Freeze Dry Flasks, Chemglass
Supplier: Chemglass
CG-1611-08 CG-1611-09 CG-1611-25 CG-1611-26 CG-1611-01 CG-1611-04 CG-1611-05 CG-1611-06 CG-1611-07 CG-1611-22 CG-1611-23 CG-1611-24
80070-444EA 563.67 CAD
80070-444 80070-446 80070-458 80070-460 80070-430 80070-436 80070-438 80070-440 80070-442 80070-452 80070-454 80070-456
Freeze Dry Flasks, Chemglass
Flasks Flat Bottom Flasks
These flat bottom borosilicate flask can be used with Labconco and Virtis freeze dryers.
Wide mouth design provides easy access sample.
Ordering information: Flasks only, not suppled with rubber top, filter or O-ring.
Caution: Labconco and Virtis are not related to or affiliated with Chemglass, and these manufacturers have not approved or validated these products for use on their equipment.
Order Now
SPECIFICATIONS
Environmentally Preferable