SystemSURE Plus™ Luminometer, ATP Hygiene Monitoring System, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
SS3 US2020
CA89492-626EA 8278.9 CAD
CA89492-626 CA10142-318
SystemSURE Plus™ Luminometer, ATP Hygiene Monitoring System, Hardy Diagnostics
Monitoring Systems Hygiene Monitoring Systems
SystemSURE Plus™ is the next-generation system of the SystemSURE II ATP hygiene monitoring system. With improved functionality, new software, and increased memory, the SystemSURE Plus meets the demands of the largest and smallest companies around the world.
  • Provides real-time feedback and facilitates long-term data analysis
  • Optimizes and verifies that cleaning procedures are working
  • Assists in developing and improving process and risk assessment programs (ie. HACCP & GMPs)
  • No scheduled yearly maintenance is necessary
  • Powered by two AA batteries