Pierce™ Nitrocellulose Membranes, Thermo Scientific
Supplier: Thermo Scientific
88014 88018 88024 88025 77010 77012 88013
CAPI88014PK 358.2 CAD
CAPI88014 CAPI88018 CAPI88024 CAPI88025 CAPI77010 CAPI77012 CAPI88013
Pierce™ Nitrocellulose Membranes, Thermo Scientific
Transfer Membranes
Nitrocellulose membranes are sheets and rolls of nitrocellulose, available in two pore sizes, that have excellent binding properties for Western blotting, dot-blot assays and other protein or nucleic acid methods.
  • Pure, 100% nitrocellulose membranes with high surface area and excellent uniformity
  • Pore size of 0.45 µM suitable for protein applications or 0.2 µM for peptides
  • Available as rolls or convenient 8x12 cm pre-cut sheets
  • High-binding affinity provides excellent protein binding, blocks easily, and produces very low background in chemiluminescent Western blotting
Nitrocellulose is a popular binding matrix for Western blotting because of its high affinity for proteins and compatibility with a variety of detection methods.
These transfer membranes have a high surface area and excellent uniformity. They are available in two sizes and suitable for either peptide or protein applications.
Nitrocellulose membranes provide excellent protein binding, block easily, and produce very low background in chemiluminescent Western blotting.