Caron's revised 7400-Series Reach-in CO2 Incubators–high-precision incubation, now available in full-size.
- Culture quality
- Clean conditions
- Facility-friendly
Caron’s Reach-in CO2 Incubators provide high precision incubation conditions, now in a full-sized package.
Get the critical growth conditions you expect from a small CO2 incubator, now in a high-capacity unit. Uniform back to front directed airflow and a tightly-sealed foam-insulated chamber produce tight ±0.3 °C temperature uniformity, even when shelves are full. Latest-generation IR CO2 sensor is highly stable, even when RH conditions vary, and drift-resistant. Optional one-handed door latch prevents temperature and humidity loss due to incomplete door closure. And the intuitive touchscreen controller helps you stay ahead of preventative maintenance situations, with easy to understand alerts.
Caron keeps the workhorse reach-in working clean. Exclusive optional 2-hour H2O2 sterilization proves fast and validatable contamination elimination, with no post-cycle wipedown or reassembly. Tool-free interior disassembles in under three minutes, for fast and easy cleanup of spills. Finally, available controlled RH system eliminates contamination-prone water pans, moving the water source outside of the growth environment.
Caron’s reach-ins are built to work on your site, whether or not your site was built to accommodate large chambers. Standard plug-in electrical service and low power usage reduce both setup and installation costs. Intuitive touchscreen controllers with breadcrumb navigation are easy to use, minimizing training time. With a choice between 25 cubic foot units that roll under a standard door or max-capacity 33 cubic foot chambers, Caron gives you options for both standard labs, as well as purpose-designed spaces. Caron even helps eliminate unforeseen costs, by including features you need, such as corrosion-resistant stainless steel interiors, glass doors, access ports, and both casters and levelling feet, as standard on every reach-in CO2 incubator.
Note: View the Product Accessories List, in Datasheets under Related Information. To order specific accessories, search by part number.
Certifications: Models 7400-25-3 and 7400-33-3 are CE compliant.
Ordering information: Accessories available separately.