Flow Control Adapters with Spherical Joints, Chemglass
Supplier: Chemglass
CG-1201-11 CG-1201-09 CG-1201-10
80067-116EA 614.33 CAD
80067-116 80067-112 80067-114
Flow Control Adapters with Spherical Joints, Chemglass
Adapters Adapters, Spherical Ground Joints
These flow control adapters are for use with Chemglass absorption chromatography columns.
  • Chem-Cap® metering plug
  • Replacement plugs available
Adapters have a Chem-Cap® metering plug. Replacement plugs are available.