Optimum Growth™ Flask
Supplier: THOMSON INSTRUMENT CO
931114 931116 931112 931113 931110 931111
76808-398CS 366.72 CAD
76808-398 76808-400 76808-394 76808-396 76808-390 76808-392
Optimum Growth™ Flask
Flasks Cell Culture Flasks
The Optimum Growth™ flasks provide excellent growth with space saving capability.
  • Baffles designed for high aeration and low shear
  • Same footprint as comparable Fernbach flask
  • Less foaming than disposable Fernbach
  • Transfer cap option connects directly to cell bags or bioreactors with quick connect, luer lock or tube fusing
  • 0.2 μm vented cap
  • Individually packaged and sterilized
They are designed in a way that protein production will scale consistently across all sizes, unlike any other shake flasks on the market. Additionally, these flask features allow for consistent shake speeds from the 125 ml up to the 7 L flasks.