Round German Glass Cover Slips, Chemglass
Supplier: Chemglass
CLS-1760-012 CLS-1760-015 CLS-1760-025
89167-106EA 239.73 CAD
89167-106 89167-108 89167-110
Round German Glass Cover Slips, Chemglass
Cover Glasses
Cover slips are manufactured from the highest quality hydrolytic Class 1 German borosilicate glass
All cover slips are #1 thickness. They have excellent resistance to chemicals and are ideal for neural tissue culture and hard-to-attach cells. These cover slips are exceptional for florescence and confocal microscopy. Sold in 1 oz. packages.