Accessories for Qsonica Sonicators
Supplier: QSonica, LLC.
432A 432B2
89207-032EA 2367.55 CAD
89207-032 89410-224
Accessories for Qsonica Sonicators
Homogenizers Ultrasonic Cell Disruptors
Sonicators are loud devices that will cause discomfort to the user and anyone nearby
The interior walls are lined with water-resistant acoustical foam and the door has a window, allowing experiments to be visually monitored. A port is located on either side of the enclosure for coolant tubing if required.
The Sound Enclosure reduces noise by approximately 20dBa, and is designed to work with all accessories. In addition to reducing noise, the Sound Enclosure has an internal support rod and converter mounting system. Any Qsonica probe or horn for the Q125/Q55 will be held safely andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable