Anti-GABPB2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20870-1-AP-150UL
10087-234EA 709.42 CAD
10087-234
Anti-GABPB2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: GABPB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR