Anti-ENOPH1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17731-1-AP-150UL
10081-292EA 709.42 CAD
10081-292
Anti-ENOPH1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HL-60 Cells, 1:500-1:5000

Type: Primary
Antigen: ENOPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR