Anti-Calpain 9 Rabbit Polyclonal Antibody

Supplier: Proteintech
17556-1-AP-150UL
10084-008EA 709.42 CAD
10084-008
Anti-Calpain 9 Rabbit Polyclonal Antibody
Antibodies
Calpain 9 is also named as NCL4 and belongs to the peptidase C2 family.It can be down-regulated in gastric cancer tissue and in gastric cell lines of differentiated and poorly differentiated types.It is predominantly expressed in the gastrointestinal tract and restricted to the gastric surface mucus (pit) cells in the stomach and the function of it is implicated in suppressing tumorigenesis.

Western Blot: Human Stomach Tissue, 1:500-1:5000; IHC: Human Stomach cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: Calpain 9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR