Anti-MBP-Tag Rabbit Polyclonal Antibody

Supplier: Proteintech
15089-1-AP
10090-022EA 693.62 CAD
10090-022
Anti-MBP-Tag Rabbit Polyclonal Antibody
Antibodies
Protein tags are protein or peptide sequences located either on the C- or N- terminal of the target protein, which facilitates one or several of the following characteristics: solubility, detection, purification, localization and expression. Maltose binding protein (MBP) is the 370 amino acid product of the E.coli mal E gene. MBP is a useful affinity tag that can increase the expression level and solubility of the resulting tagged protein. The MBP tag also promotes proper folding of the attached protein. Plasmid vectors have been constructed utilizing the MBP domain that allow the synthesis of high levels of MBP-fusion proteins that can be purified in a one step procedure by affinity chromatography cross linked amylose resin. Once bound to amylose, the MBP protein can then be separated from the target protein by cleavage by coagulation Factor Xa at a specific four residue site. Alternatively, the intact fusion protein can be specifically eluted from the resin by the addition of excess free maltose. Subsequent to elution, MBP fusion protein can be visualized either by western blot analysis or immunoprecipitation using antibodies specific for the MBP-tag. This antibody recognizes MBP (Maltose binding protein) TAG in some expression systems.

Western Blot:lane1: MBP-NOL6 64kd; lane2 MBP 42kd, 1:1000-1:10000

Type: Primary
Antigen: MBP-Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR