Anti-DNM3 Rabbit Polyclonal Antibody

Supplier: Proteintech
14737-1-AP
10086-044EA 709.42 CAD
10086-044
Anti-DNM3 Rabbit Polyclonal Antibody
Antibodies
DNM3 (dynamin 3) is a member of dynamin proteins that participate in membrane rearrangements such as vesicular trafficking and phaocytosis. Three dynamin proteins encoded by different genes are present in mammals: DNM1 is mainly expressed in central nervous system, DNM2 ubiquitously expressed, and DNM3 expressed chiefly in the testis and brain. Western blot using this antibody specifically detected the 100 kDa DNM3 in mouse brain. It doesn't cross-react with other DNMs. (22977254, 21671749)

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: DNM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR