Anti-COX6B1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11425-1-AP-150UL
10085-068EA 709.42 CAD
10085-068
Anti-COX6B1 Rabbit Polyclonal Antibody
Antibodies
COX6B1 (Cytochrome c oxidase subunit 6B1) is also named as COX6B and belongs to the cytochrome c oxidase subunit 6B family. It connects the two COX monomers into the physiological dimeric form.Defects in COX6B1 are a cause of mitochondrial complex IV deficiency (MT-C4D).

Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: COX6B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR