Anti-USP11 Rabbit Polyclonal Antibody

Supplier: Proteintech
10244-1-AP-150UL
10082-242EA 709.42 CAD
10082-242
Anti-USP11 Rabbit Polyclonal Antibody
Antibodies
USP11(Ubiquitin carboxyl-terminal hydrolase 11), also named as UHX1, is a deubiquitinating enzyme controling many intracellular processes, including cell cycle progression, transcriptional activation, and signal transduction. There are four bands of 120 kDa, 55 kDa, 50 kDa and 44 kDa to be detected in HeLa extract by western blot and the smaller proteins may be degradation products, some other spliced products could possibly correspond to the smaller proteins and possess functions. This protein belongs to the peptidase C19 family.

Western Blot: Human Testis Tissue, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: USP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR