Axygen® Robotic Pipette Tips for AP96, AP384 and FX/NX Series Platforms, Corning
Supplier: Corning
FX-384-XL-R FX-384-XL-R-S FXF-384-XL-R-S FX-255-WB-R FX-384-R FX-384-R-S FXF-165-WB-R-S FX-10-R FX-384-30FPR FXF-180-R-PB FX-20-R FX-20-R-S FXF-20-R-S FX-50-R FX-50-R-S FXF-50-R-S FX-250-R FX-250-R-S FXF-180-R-S FX-20-L-R FXF-20-L-R-S FX-250-L-R-S FXF-200-R-S FX-384-96-R FX-384-XL-R FX-384-XL-R-S
12000-194CS 4677.68 CAD
12000-194 12000-196 12000-198 12000-200 12000-204 12000-696 47751-032 80078-410 80078-414 80078-416 80078-418 80078-820 89005-646 89125-002 89204-790 89204-792 89204-794 89204-796 89204-798 89204-800 89204-802 89204-804 89204-806 89204-808 89204-812 89204-816 89204-818 89204-822 89204-824 89204-826 CA10754-534 CA76553-880 CA76553-878
Axygen® Robotic Pipette Tips for AP96, AP384 and FX/NX Series Platforms, Corning
Pipette Tips
Free of RNase, DNase, DNA and pyrogens.
  • Transparent PP
  • Sterile tips: Sterilised by gamma radiation
  • Filter tips: PE filter prevents any contamination of the pipette
  • Maxymum Recovery™ tips: Ultra-smooth surface prevents retention, loss of components, and denaturation of the sample