Pearl® Adjustable Volume Micropipettes, 8-Channel
Supplier: Heathrow Scientific
HS120215 HS120216
10127-198EA 1402.91 CAD
10127-198 10127-200
Pearl® Adjustable Volume Micropipettes, 8-Channel
Pipettes Micropipettes
Pearl® adjustable volume 8-channel micro pipettors feature a sleek erogonomic design and reduced plunger pressure to help reduce the chance of RSI, as the pipettes require ²/₃rd the operation pressure of most standard pipettes.
  • Volume adjustment and low-force dispensing all in one button for convenience
  • Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
  • Isolated volume locking ring prevents accidental volume changes
  • Finger hook
  • 3-position volume display
  • Universal tip cone
  • Autoclavable
All devices feature internal metal parts for increased accuracy and calabiration stability, easy volume adjustment with locking tab to prevent accidental change, and separate tip ejector and a large digital volume display.
Each pipette is tested in accordance with standards and a calibration certificate is supplied.
Certifications: CE, UKCA.
Delivery information: Supplied with one year warrenty.