Absorption Chromatography Columns, Chemglass
Supplier: Chemglass
CG-1198-39 CG-1198-40 CG-1198-44 CG-1198-45 CG-1198-49 CG-1198-50 CG-1198-22 CG-1198-23 CG-1198-26 CG-1198-27 CG-1198-30 CG-1198-31 CG-1198-34 CG-1198-35
80067-014EA 784.86 CAD
80067-014 80067-016 80067-022 80067-024 80067-030 80067-032 80066-980 80066-982 80066-988 80066-990 80066-996 80066-998 80067-004 80067-006
Absorption Chromatography Columns, Chemglass
Chromatography Columns
Absorption chromatography columns are useful for rapid preparative separations.
Moderate resolution can be obtained with this inexpensive to setup and operate column. Columns are constructed using MEDIUM WALL tubing. Glass wool (not supplied) is used to support column packing. Columns can be used with optional flow controller and reservoir. The connections between the column, reservoir and flow control adapter are spherical joints connected with stainless steel clamps.