NP-40 Alternative, MilliporeSigma
Supplier: MilliporeSigma
492016-100ML 492016-500ML
CA80601-882EA 212.22 CAD
CA80601-882 CA80601-884
NP-40 Alternative, MilliporeSigma
Protein Extraction Protein Extraction Reagents
NP-40 alternative is a non-ionic surfactant used in the isolation and purification of functional membrane protein complexes and in two-dimensional (2D) gel electrophoresis.