Whatman™ Specialized Test Papers
Supplier: Cytiva
2602-500A 2602-501A 2651-500 2651-500 2602-500A 2602-501A
14217-692PK 80.72 CAD
14217-692 14217-694 CA11013-716 10035-368 CA14217-692 CA14217-694
Whatman™ Specialized Test Papers
Test Papers Indicator Papers
Cytiva offers a range of pH indicator and test papers to meet specific needs.
  • Convenient and portable for field use
Potassium iodide test paper: For detecting chlorine and other oxidising agents. In acid solution, oxidising agents react with the iodide in the test paper to liberate iodine. The paper turns blue in the presence of an oxidizing agent (e.g., CI₂, Br₂, H₂O₂, HNO₂ etc.).
Lead acetate test paper: Used for detecting hydrogen sulfide, this rapid qualitative test paper when wetted with distilled water can detect as little as 5 ppm of H₂S inMore Product Information