Wescorp ESD High Temperature Polymide Tape, Desco
Supplier: Desco
81271 81270
47732-144EA 68.7 CAD
47732-144 47732-146
Wescorp ESD High Temperature Polymide Tape, Desco
Tapes
Used in applications masking PCB gold features for wave soldering and IR reflow ovens
  • Removal leaves little or no residue
  • Static charge generation during unwind and peel off stainless steel plate application is 5 volts @ 50% RH
  • 1.0 mil thick (0.001" or 0.0254 mm) Polyimide Film (DuPont Kapton® or equivalent)
  • 1.4 mil thick (0.0014" or 0.0356 mm) conductive polysiloxide adhesive
  • Adhesive surface resistivity 10E3 to 10E4 ohms
  • Max Temperature 260°C (500°F)
  • For best results, apply to board using a rubber roller
Tapes should be stored in a dry, wellMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable