TLC Sprayers and Components, Chemglass
Supplier: Chemglass
CG-1180-02 CG-1180-03 CG-1180-04 CG-1180-06 CG-1180-07 CG-1180-08 CG-1180-10 CG-1180-11 CG-1180-12 CG-182-03
80060-588EA 493.66 CAD
80060-588 80060-590 80060-592 80060-594 80060-596 80060-598 80060-600 80060-602 80060-604 80073-400
TLC Sprayers and Components, Chemglass
Atomizers
These TLC sprayers provide a fine, uniform spray ideally adjusted for the development of chromatographic patterns
Spray pattern is adjustable by varying the coverage of the vent hole with the user’s thumb. Sprayer is used with low pressure air (5/PSI) or rubber pressure bulb. Sprayer consists of an atomizer top attached to the reservoir with a Rodaviss® 24/40 joint. This design provides for a positive, grease-free seal between the atomizer and reservoir and eliminates the glass hooks used on other sprayers. Complete sprayer consists of the atomizer top, Erlenmeyer shaped reservoir, open-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable