SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: Eppendorf
5361000031 5360000038 5362000035 5366000021 5365000028 5364000024 5363000039 5306000006 5307000000 5309000007 5310000002 5316000004 5318000001
CA11028-282EA 1465.11 CAD
CA11028-282 CA11028-284 CA11028-286 CA11028-288 CA11028-290 CA11028-292 CA11028-294 CA11028-296 CA11028-298 CA11028-300 CA11028-302 CA75856-522 CA75856-524 CA77321-626
SmartBlocks for ThermoMixer® C and ThermoStat C
Block Heaters Blocks
Exchangeable in just seconds. Individually sensor-controlled SmartBlocks are your solution for maintaining accurate, uniform temperatures. A wide variety of these flexible thermoblocks for all common laboratory vessels support a broad range of applications - for use with Eppendorf ThermoMixer® C and ThermoStat C.
  • Optimized block design supports optimal temperature transfer to sample to achieve excellent results
  • Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
  • Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept®)
  • Automatic block recognition for safe handling
  • Optimized block design ensures maximum temperature transfer to the sample
  • Eppendorf QuickRelease technology enables fast and simple block exchange – no need for tools
Every Eppendorf SmartBlock is individually inspected in production for temperature accuracy and includes a serial-number specific certificate for documentation purpose.