Magnetic Stirring Bars, Octagonal,PTFE, Chemglass
Supplier: Chemglass
CG-2001-50 CG-2001-01 CG-2001-02 CG-2001-03 CG-2001-04 CG-2001-06 CG-2001-07 CG-2001-08 CG-2001-09 CG-2001-10 CG-2001-11 CG-2001-12 CG-2001-13 CG-2001-15 CG-2001-17 CG-2001-18 CG-2001-19 CG-2001-20
89028-972EA 435.55 CAD
89028-972 80062-040 80062-042 80062-044 80062-046 80062-048 80062-050 80062-052 80062-054 80062-056 80062-058 80062-060 80062-062 80062-064 80062-066 80062-068 80062-070 80062-072 80062-074 80062-076 80062-078
Magnetic Stirring Bars, Octagonal,PTFE, Chemglass
Stirrers Magnetic Stirring Bars
Octagon cross-section features a molded-in pivot ring
The PTFE coated Alnico magnet permits magnetic agitation without contamination.