DISPOSAFLEX® Glass Columns, Kimble Chase
Supplier: DWK Life Sciences
420166-1001 420166-0001
KT420166-0001PK 1399.78 CAD
KT420166-0001 KT420166-1001
DISPOSAFLEX® Glass Columns, Kimble Chase
Chromatography Columns
A multi-purpose, disposable glass column.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Supplied assembled with a polypropylene reservoir, a lower column fitting with a luer outlet and a 20 µm polyethylene bed support.