Compact Digital Overhead Stirrers, Caframo
Supplier: Caframo
BDC2002 BDC2002U1
CA58960-174EA 1720.42 CAD
CA58960-174 CA10860-014
Compact Digital Overhead Stirrers, Caframo
Stirrers Overhead Stirrers
This Compact Digital Overhead Stirrer maximizes productivity, fits into any area of the laboratory including a fume hood or enclosed workstation, and provides accurate and repeatable results every time.
Suitable for a wide range of mixing applications, this Caframo benchtop overhead stirrers is used in formulations, reaction systems, pharmaceuticals, QA testing, and education classrooms.
With the push of a button, the unique Stirlight™ lights up so that solutions can be seen clearly. The Compact Digital stirrer is powered by a brushless DC (BDC) motor. The belt driven transmission isMore Product Information