VWR® Paper Weigh Boats
Supplier: VWR International
10048-874 10048-950 10048-952 10048-954 10755-254 10816-244
10048-874CS 858.36 CAD
10048-874 10048-950 10048-952 10048-954 10755-254 10816-244
VWR® Paper Weigh Boats
Boats Weighing Boats
A simple and cost-effective solution to reduce plastic use in the lab, VWR® Paper Weighing Boats are both Eco-IN and Eco-OUT.
  • Grease-proof paper construction
  • Naturally biodegradable
  • Control contents like plastic weigh boats
They are made of FSC® certified paper (Eco-IN) and will naturally biodegrade in the standard municipal waste stream (Eco-OUT)
Made of grease-proof paper that meets FDA 21 CFR 176.170 (components of paper in contact with aqueous and fatty foods), and with 2I CFR 176.180 (contact with dry foods). Compatible with heat applications with a temperature limitation of 210°C (410°F).
Available in small and medium sizes, in either brown for light colored samples or white for dark samples. Medium boats are made in USA, small and large boats are made in Italy.
VWR® Paper Weigh Boats control contents like plastic weigh boats and are easier to handle than weighing paper.
Certifications: USA: FSC®Certifed Paper, ISO 9002 certified facility in accordance with BGvv, Haccp, and FDA regulations. FSC Certification. EU: FSC®Certifed Paper, ISO 14001 Certification - Environmental policy, BRC/IOP Certification, ISO 9001 Certification.