Adventurer® AX Precision Balances,Ohaus®
Supplier: Ohaus
30100606 30100605 30100608 30100610 30100609 30100612 30100611 30100614 30100613 30100616 30100615 30100617 30100618 30100631 30100632 30100633 30100634 30100635 30100636 30100637 30122655 30100639 30122635 30122634 30100619 30127981 30127983 30248912
10153-704EA 3222.84 CAD
10153-704 10153-706 10153-708 10153-712 10153-714 10153-720 10153-722 10153-724 10153-726 10153-728 10153-730 10153-732 10153-734 10153-736 10153-738 10153-740 10153-742 10153-744 10153-746 10153-748 10153-750 10153-752 10153-768 10153-770 10153-772 10820-642 10820-644 10820-646
Adventurer® AX Precision Balances,Ohaus®
Balances and Scales
The Adventurer® AX Precision balance series strikes the ideal mix between inventive features, long-lasting durability, and uncomplicated weighing capabilities
The Adventurer® AX incorporates all of the features and applications necessary for simple weighing and also offers options for more complex measurements. Quick and easy weighing are possible from a large color touchscreen display, extremely fast stabilization times, GLP/GMP compliance, two USB ports, ingenious draftshield on 1mg models, multi-language software, and much more. The Adventurer® AX is the most complete and user friendlyMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable