Adventurer® AX Analytical Balances, Ohaus®
Supplier: Ohaus
30100601 30100600 30100603 30100602 30100604 30100630
10153-692EA 5832.27 CAD
10153-692 10153-694 10153-696 10153-698 10153-700 10153-702
Adventurer® AX Analytical Balances, Ohaus®
Balances and Scales
The Adventurer® AX Analytical balance series strike the ideal mix between inventive features, long-lasting durability, and uncomplicated weighing capabilities.
The Adventurer® AX incorporates all of the features and applications necessary for simple weighing and also offers options for more complex measurements
Stability, accuracy, and fast operation ensure optimal weighing results. This trio of characteristics ensures the most important aspects of routine laboratory weighing are covered: accurate results you can count on extremely quickly. The color touchscreen and connectivity options power a modern weighing experience. The wide viewing angle color touchscreen provides quick access to all of the unit’s applications, while dual USB ports with GLP/GMP tracking capabilities assist in the monitoring and reporting of data. A space-saving draftshield improves user experience and accessibility. The two-piece, top-mounted draftshield (mg and 0.1mg models only) doors open wide on both sides, providing ample room for sample placement. This also reduces the footprint of the draftshield in the rear of the balance when open.
Operate and access the nine application modes and abundant features that eliminate the need to do several manual calculations through the modern color touchscreen. Operators can wear laboratory gloves while utilizing the touchscreen, eliminating the inconvenience and hazards associated with constantly putting on and removing gloves. In addition to the touchscreen, the Adventurer® AX also features six mechanical keys that provide tactile feedback and allow the operator to perform repetitive operations such as tare, zero, calibration, and print. All units feature a die-cast metal bottom housing, stainless steel pan, full housing replaceable in-use cover, and integral weigh below hook.
Applications include Weighing, Parts Counting, Percent Weighing, Check Weighing, Animal/Dynamic Weighing, Formulation, Totalization, Density Determination, and Display Hold.
Certifications: Metrology: NIST Handbook 44, Measurement Canada Weights and Measures Regulations (Class I, nmax 220000; Class II, nmax 52000). Product Safety: CSA C22.2 No. 61010-1, UL 61010-1, IEC 61010-1. Electromagnetic Compatibility: FCC Part 15 Class A, ICES-001 Class A, IEC 61326-1 (emissions Class B, immunity Industrial requirements).
Ordering information: Adventurer® AX balances can be purchased with or without legal for trade capability, and with or without internal calibration depending on model. AC adapter included.