Calcium oxide 95,0 Complexometric, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime

MQ200

1.02109.1000 1.02109.9025
CA1.02109.1000CS 2636 CAD
CA1.02109.1000 CA1.02109.9025
Calcium oxide 95,0 Complexometric, Supelco®
Calcium oxide
Formula: CaO
MW: 56.08 g/mol
Boiling Pt: 2850
Melting Pt: 2580
Density: 3.37 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
UN: 1910
Merck Index: 13,01691

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR