Life/form® Complete Auscultation Training Set
Supplier: Nasco
LF01144 LF01145 LF01190 LF01191 LF01142
470119-470EA 3255.2 CAD
470119-470 470119-472 470120-414 470120-416 470125-148
Life/form® Complete Auscultation Training Set
Educational Materials Biology Educational Materials Human Biology Learning Activities
Student And Patient Training Tools
  • Combine the board and torsos for complete training
  • Board folds for easy storage and transport
  • Hear 16 different heart and lung sounds
The instructor selects from a menu of heart and lung conditions by wireless remote control. The manikin presents itself as a real patient without visible auscultation sites. The student must palpate to identify correct auscultation locations, and will hear different heart and lung sounds as the SmartScope™ is moved to different locations on the manikin. A diagnosis of the condition selected by the instructor can be made by comparing the variations in sounds occurring at different sites. Just like a real patient! Lung sounds can be detected at five anterior, 10 posterior, and two midaxillary locations. Students can practice auscultation at six anterior heart sites. The remote control does not have to be pointed directly at the manikin or SmartScope™ to operate. One remote control will operate all SmartScope™ within a 100 ft. range. Great for group instruction. Listen to the sounds by either using headpieces on SmartScope™ or by connecting to an amplifier/speaker (not included). Range on the unit is up to 100 ft. of remote access.
Complete Set includes Board and case, manikin, one remote control with LCD display, one SmartScope™ with single- and dual-user headpieces, and hard carry case. Operates using two “AA” and two “AAA” batteries (included).
The Auscultation Package includes all of the above, except for the Board and Case.
Lung Conditions to Select with the Remote: Bronchovesicular, Bronchial, Cavernous, Coarse Crackle, Egophony, Fine Crackle, Friction Rub, Infant, Mono Wheeze, Normal Lung, Pectoriloquy, Pulmonary Edema, Rhonch, Stridor, Vesicular, and Wheeze. Heart Conditions to Select with Remote: Aortic Regurgitation, Atrial Septal Defect, Holosystolic, Midsystolic, Mitral Stenosis, Normal, PDA, Pulmonary, Stenosis, S3 Gallop, S4 Gallop, Systolic Click, and VSD.