Granular dessicant, Sicapent® with indicator, MilliporeSigma
Supplier: MilliporeSigma
1.00543.2800 1.00543.2800 1.00543.0500 1.00543.0500
CA1.00543.0500CS 2531.51 CAD
CA1.00543.0500 CA1.00543.2800 EM1.00543.2800 EM1.00543.0500
Granular dessicant, Sicapent® with indicator, MilliporeSigma
Desiccators Desiccants
Sicapent is a granular dessicant comprising ~75% phoshorus pentoxide on an inert inorganic carrier.
  • Ease of use
  • Fast water adsorption due to large surface area
  • Colour indicator - colourless to green to blue
  • Remains in particle form after 100% water uptake
1.00543.2800 replaces and is the same as SX0137-1. Part # Change only, Same Pack Size, Same Specifications.
Appliations include drying liquids, filling drying tubes, particularly for fast flowing gases in drying tubes
Bulk density: ~300 g/l