Measuring Cylinders, To Contain, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
20027-10 20027-25 20027-50 20027-100 20027-250 20027-500 20027-1000 20027-2000
89217-664CS 443.49 CAD
89217-664 89217-666 89217-668 89217-670 89217-672 89217-674 89217-676 89217-678
Measuring Cylinders, To Contain, Class A, Kimble Chase
Cylinders Measuring Cylinders
Class A, to contain, measuring cylinders feature a pour spot top
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: A SAFE-GARD® bumper is supplied with each measuring cylinder.