VWR® Heavy-Duty Wide Mouth Volumetric Flasks with PTFE Stoppers, Amber Glass, Class A, Serialized
Supplier: VWR International
10124-096 10124-098 10124-100 10124-102 10124-104 10124-106 10124-192 10124-194
10124-096CS 1078.32 CAD
10124-096 10124-098 10124-100 10124-102 10124-104 10124-106 10124-192 10124-194
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with PTFE Stoppers, Amber Glass, Class A, Serialized
Flasks Volumetric Flasks
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
  • Borosilicate glass fabrication
  • Flasks are individually serialized
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with PTFE stopper.
Ordering information: Flasks come with individual certificate of compliance.