VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Amber Glass, Class A, Serialized
Supplier: VWR International
10124-086 10124-088 10124-090 10124-092 10124-094 10124-172 10124-174 10124-176
10124-086CS 875.06 CAD
10124-086 10124-088 10124-090 10124-092 10124-094 10124-172 10124-174 10124-176
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Amber Glass, Class A, Serialized
Flasks Volumetric Flasks
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
  • Borosilicate glass fabrication
  • Flasks are individually serialized
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with glass penny head stopper.
Ordering information: Flasks come with individual certificate of compliance.