Purospher® STAR RP-18 Endcapped UHPLC Columns, MilliporeSigma
Supplier: MilliporeSigma
Purospher® STAR, Hibar®
1.50646.0001 1.50653.0001 1.50650.0001 1.50651.0001 1.50649.0001 1.50648.0001 1.50654.0001 1.50651.0001 1.50650.0001 1.50646.0001 1.50649.0001 1.50648.0001 1.50653.0001 1.50645.0001 1.50645.0001 1.50654.0001
CA1.50645.0001EA 2567.7 CAD
CA1.50645.0001 EM1.50649.0001 CA1.50649.0001 97021-982 97021-980 CA1.50650.0001 CA1.50646.0001 97021-985 CA97021-984 CA97021-983 97021-986 97021-983 97021-984 97021-978 CA97021-986 CA97021-985
Purospher® STAR RP-18 Endcapped UHPLC Columns, MilliporeSigma
Chromatography Columns
Available with optimised 2 μm particulate silica, these UHPLC columns based on Purospher® STAR RP-18 endcapped are ideal for ultra-fast applications, where resolution, sensitivity and sample throughput are crucial.
  • In UHPLC environment, speed is increased by a factor of 10
  • Solvent consumption is drastically cut
  • Excellent peak shape for all types of acidic, basic and metal chelating analytes
  • Extraordinary pH stability from pH 1.5 to 10.5 for an extremely wide application range
  • Enhanced sensitivity due to improved signal-to-noise ratio
Due to its balanced selectivity, Purospher® STAR RP-18 endcapped UHPLC columns cover almost all demanding separations with tailing-free chromatograms. The Purospher® STAR RP-18 endcapped 3 μm columns are recommended for difficult samples where clogging and back pressure present an issue.
Hibar® HR columns are complete with end fittings and pressure stable (600 bar) for UHPLC use.
They are the first choice for high-throughput screening and QC analyses, process monitoring, method development, and LC/MS applications.